MilliporeSigma
Single beam low frequency 2D Raman spectroscopy.
Optics express (2020-03-04)
Ilan Hurwitz, Dekel Raanan, Liqing Ren, Hadas Frostig, Patric Oulevey, Barry D Bruner, Nirit Dudovich, Yaron Silberberg
ABSTRACT
Low frequency Raman spectroscopy resolves the slow vibrations resulting from collective motions of molecular structures. This frequency region is extremely challenging to access via other multidimensional methods such as 2D-IR. In this paper, we describe a new scheme which measures 2D Raman spectra in the low frequency regime. We separate the pulse into a spectrally shaped pump and a transform-limited probe, which can be distinguished by their polarization states. Low frequency 2D Raman spectra in liquid tetrabromoethane are presented, revealing coupling dynamics at frequencies as low as 115 cm-1. The experimental results are supported by numerical simulations which replicate the key features of the measurement. This method opens the door for the deeper exploration of vibrational energy surfaces in complex molecular structures.
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, purum p.a., for separation of mineral compounds, ≥98.0% (GC)