MilliporeSigma
  • Self-assembly Synthesis of Molecularly Imprinted Polymers for the Ultrasensitive Electrochemical Determination of Testosterone.
Self-assembly Synthesis of Molecularly Imprinted Polymers for the Ultrasensitive Electrochemical Determination of Testosterone.
Biosensors (2020-03-04)
Kai-Hsi Liu, Danny O'Hare, James L Thomas, Han-Zhang Guo, Chien-Hsin Yang, Mei-Hwa Lee
ABSTRACT
Molecularly imprinted polymers (MIPs) can often bind target molecules with high selectivity and specificity. When used as MIPs, conductive polymers may have unique binding capabilities; they often contain aromatic rings and functional groups, which can undergo π-π and hydrogen bonding interactions with similarly structured target (or template) molecules. In this work, an electrochemical method was used to optimize the synthetic self-assembly of poly(aniline-co-metanilic acid) and testosterone, forming testosterone-imprinted electronically conductive polymers (TIECPs) on sensing electrodes. The linear sensing range for testosterone was from 0.1 to 100 pg/mL, and the limit of detection was as low as ~pM. Random urine samples were collected and diluted 1000-fold to measure testosterone concentration using the above TIECP sensors; results were compared with a commercial ARCHITECT ci 8200 system. The testosterone concentrations in the tested samples were in the range of 0.33 ± 0.09 to 9.13 ± 1.33 ng/mL. The mean accuracy of the TIECP-coated sensors was 90.3 ± 7.0%.
Sigma-Aldrich
3-Aminobenzenesulfonic acid, 97%