MilliporeSigma
  • Spontaneous self-intercalation of copper atoms into transition metal dichalcogenides.
Spontaneous self-intercalation of copper atoms into transition metal dichalcogenides.
Science advances (2020-02-29)
Xiao-Chen Liu, Shuyang Zhao, Xueping Sun, Liangzi Deng, Xiaolong Zou, Youcheng Hu, Yun-Xiao Wang, Ching-Wu Chu, Jia Li, Jingjie Wu, Fu-Sheng Ke, Pulickel M Ajayan
ABSTRACT
Intercalated transition metal dichalcogenides (TMDs) have attracted substantial interest due to their exciting electronic properties. Here, we report a unique approach where copper (Cu) atoms from bulk Cu solid intercalate spontaneously into van der Waals (vdW) gaps of group IV and V layered TMDs at room temperature and atmospheric pressure. This distinctive phenomenon is used to develop a strategy to synthesize Cu species-intercalated layered TMD compounds. A series of Cu-intercalated 2H-NbS2 compounds were obtained with homogeneous distribution of Cu intercalates in the form of monovalent Cu (I), occupying the tetrahedral sites coordinated by S atoms within the interlayer space of NbS2. The Fermi level of NbS2 shifts up because of the intercalation of Cu, resulting in the improvement of electrical conductivity in the z-direction. On the other hand, intercalation of Cu into vdW gaps of NbS2 systematically suppresses the superconducting transition temperature (Tc) and superconducting volume fraction.
Sigma-Aldrich
Sulfur, purum p.a., ≥99% (T)