MilliporeSigma
  • Beta-Catenin Causes Adrenal Hyperplasia by Blocking Zonal Transdifferentiation.
Beta-Catenin Causes Adrenal Hyperplasia by Blocking Zonal Transdifferentiation.
Cell reports (2020-04-23)
Emanuele Pignatti, Sining Leng, Yixing Yuchi, Kleiton S Borges, Nick A Guagliardo, Manasvi S Shah, Gerard Ruiz-Babot, Dulanjalee Kariyawasam, Makoto Mark Taketo, Ji Miao, Paula Q Barrett, Diana L Carlone, David T Breault
ABSTRACT
Activating mutations in the canonical Wnt/β-catenin pathway are key drivers of hyperplasia, the gateway for tumor development. In a wide range of tissues, this occurs primarily through enhanced effects on cellular proliferation. Whether additional mechanisms contribute to β-catenin-driven hyperplasia remains unknown. The adrenal cortex is an ideal system in which to explore this question, as it undergoes hyperplasia following somatic β-catenin gain-of-function (βcat-GOF) mutations. Targeting βcat-GOF to zona Glomerulosa (zG) cells leads to a progressive hyperplastic expansion in the absence of increased proliferation. Instead, we find that hyperplasia results from a functional block in the ability of zG cells to transdifferentiate into zona Fasciculata (zF) cells. Mechanistically, zG cells demonstrate an upregulation of Pde2a, an inhibitor of zF-specific cAMP/PKA signaling. Hyperplasia is further exacerbated by trophic factor stimulation leading to organomegaly. Together, these data indicate that β-catenin drives adrenal hyperplasia through both proliferation-dependent and -independent mechanisms.
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Renin Substrate Tetradecapeptide porcine, ≥97% (HPLC)