MilliporeSigma
  • Synthesis and Properties of Thermosets from Tung Oil and Furfuryl Methacrylate.
Synthesis and Properties of Thermosets from Tung Oil and Furfuryl Methacrylate.
Polymers (2020-01-26)
Sunanda Sain, Dan Åkesson, Mikael Skrifvars
ABSTRACT
This work focuses on the development of cross-linked polymer from a highly unsaturated vegetable oil, tung oil (TO) and a bio-based acrylate, furfuryl methacrylate (FMA). The presence of a high degree of unsaturated carbon-carbon bonding in TO makes it a suitable precursor for polymer synthesis. Using this advantage of TO, in this work, we have synthesised a cross-linked polymer from TO and FMA through free radical polymerisation followed by Diels-Alder (DA) reaction. Successful incorporation of both of the raw materials and the two chemical reactions was shown using Fourier-transform infrared (FTIR) and Raman spectroscopy. The development of cross-linked structure was analysed through thermogravimetric analysis (TGA) and dynamic mechanical analysis (DMA).
Sigma-Aldrich
Furfuryl methacrylate, 97%, contains 200 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.