MilliporeSigma
  • Location, location, location: new insights into O-GalNAc protein glycosylation.
Location, location, location: new insights into O-GalNAc protein glycosylation.
Trends in cell biology (2010-12-15)
David J Gill, Henrik Clausen, Frederic Bard
ABSTRACT
O-GalNAc glycosylation of proteins confers essential structural, protective and signaling roles in eumetazoans. Addition of O-glycans onto proteins is an extremely complex process that regulates both sites of attachment and the types of oligosaccharides added. Twenty distinct polypeptide GalNAc-transferases (GalNAc-Ts) initiate O-glycosylation and fine-tuning their expression provides a mechanism for regulating this action. Recently, a new mode of regulation has emerged where activation of Src kinase selectively redistributes Golgi-localized GalNAc-Ts to the ER. This relocalization results in a strong increase in the density of O-glycan decoration. In this review, we discuss how different mechanisms can regulate the number and the types of O-glycans decorating proteins. In addition, we speculate how Src-dependent relocation of GalNAc-Ts could play an important role in cancerous cellular transformation.
Sigma-Aldrich
N-Acetyl-D-galactosamine, ≥98% (HPLC)
Pricing and availability is not currently available.