MilliporeSigma
  • Nitrate removal from aqueous solutions by magnetic cationic hydrogel: Effect of electrostatic adsorption and mechanism.
Nitrate removal from aqueous solutions by magnetic cationic hydrogel: Effect of electrostatic adsorption and mechanism.
Journal of environmental sciences (China) (2020-03-17)
Junyi Li, Shuoxun Dong, Yili Wang, Xiaomin Dou, Haotian Hao
ABSTRACT
Excessive nitrate (NO3-) is among the most problematic surface water and groundwater pollutants. In this study, a type of magnetic cationic hydrogel (MCH) is employed for NO3- adsorption and well characterized herein. Its adsorption capacity is considerably pH-dependent and achieves the optimal adsorption (maximum NO3--adsorption capacity is 95.88 ± 1.24 mg/g) when the pH level is 5.2-8.8. The fitting result using the homogeneous surface diffusion model indicates that the surface/film diffusion controls the adsorption rate, and NO3- approaches the center of MCH particles within 30 min. The diffusion coefficient (Ds) and external mass transfer coefficient (kF) in the liquid phase are 1.15 × 10-6 cm2/min and 4.5 × 10-6 cm/min, respectively. The MCH is employed to treat surface water that contains 10 mg/L of NO3-, and it is found that the optimal magnetic separation time is 1.6 min. The high-efficiency mass transfer and magnetic separation of MCH during the adsorption-regeneration process favors its application in surface water treatment. Furthermore, the study of the mechanism involved reveals that both -N+(CH3)3 groups and NO3- are convoluted in adsorption via electrostatic interactions. It is further found that ion exchange between NO3- and chlorine occurs.
Sigma-Aldrich
(3-Acrylamidopropyl)trimethylammonium chloride solution, 75 wt. % in H2O