MilliporeSigma
  • Domestic boiling and salad preparation habits affect glucosinolate degradation in red cabbage (Brassica oleracea var. capitata f. rubra).
Domestic boiling and salad preparation habits affect glucosinolate degradation in red cabbage (Brassica oleracea var. capitata f. rubra).
Food chemistry (2020-04-04)
Franziska S Hanschen
ABSTRACT
Red cabbage contains glucosinolates, precursors to health-promoting isothiocyanates. However, raw cabbage often releases mainly epithionitriles and nitriles from glucosinolates. To increase isothiocyanate formation, the effect of acid usage in the preparation of red cabbage was evaluated. Moreover, the effects of the chosen boiling method (acidic boiled red cabbage versus neutral boiled blue cabbage) on glucosinolate degradation were investigated using UHPLC-DAD-ToF-MS and GC-MS. The addition of vinegar significantly increased isothiocyanate formation of cabbage salad from 0.09 to 0.21 µmol/g fresh weight, while lemon juice only slightly increased isothiocyanate formation. Acidic boiled red cabbage degraded glucosinolates and increased nitrile formation, while in neutral boiled blue cabbage, glucosinolates were stable. However, shortly boiled blue cabbage (5 min) had the highest isothiocyanate levels (0.08 µmol/g fresh weight). Thus, for a diet rich in isothiocyanates it is recommended to acidify raw cabbage salads and prepare shortly boiled blue cabbage instead of red cabbage.
Product Description
Supelco
Benzonitrile, suitable for HPLC, 99.9%
Sigma-Aldrich
3-Phenylpropionitrile, 99%
Sigma-Aldrich
Allyl cyanide, 98%