MilliporeSigma
  • Red aggregation-induced emission luminogen and Gd3+ codoped mesoporous silica nanoparticles as dual-mode probes for fluorescent and magnetic resonance imaging.
Red aggregation-induced emission luminogen and Gd3+ codoped mesoporous silica nanoparticles as dual-mode probes for fluorescent and magnetic resonance imaging.
Journal of colloid and interface science (2020-02-12)
Ziyang He, Ruming Jiang, Wei Long, Hongye Huang, Meiying Liu, Yulin Feng, Naigen Zhou, Hui Ouyang, Xiaoyong Zhang, Yen Wei
ABSTRACT
Fluorescence imaging and magnetic resonance imaging have been research hotspots for adjuvant therapy and diagnosis. However, traditional fluorescent probes or contrast agents possess insurmountable weaknesses. In this work, we reported the preparation of dual-mode probes based on mesoporous silica nanomaterials (MSNs), which were doped with an aggregation-induced emission (AIE) dye and Gd3+ through a direct sol-gel method. In this system, the obtained materials emitted strong red fluorescence, in which the maximum emission wavelength was located at 669 nm, and could be applied as effective fluorescence probes for fluorescence microscopy imaging. Furthermore, the introduction of Gd3+ made the nanoparticles effective contrast agents when applied in contrast-enhanced magnetic resonance (MR) imaging because they could improve the contrast of MR imaging. The excellent biocompatibility of these nanoparticles, as demonstrated via a typical CCK-8 assay, and their performance in fluorescence cell imaging and MR imaging shows their potential for applications in biomedical imaging.
Sigma-Aldrich
4-(Diphenylamino)phenylboronic acid, ≥95%