MilliporeSigma
  • Construction of a novel Staphylokinase (SAK) mutant with low immunogenicity and its evaluation in rhesus monkey.
Construction of a novel Staphylokinase (SAK) mutant with low immunogenicity and its evaluation in rhesus monkey.
International journal of biological macromolecules (2019-11-16)
Min Wang, Yao Chen, Wenliang Fu, Minji Zou, Yuanyuan Wang, Weiwei Xing, Jiaxi Wang, Donggang Xu
ABSTRACT
The heterologous nature of SAK, a thrombolytic drug, elicits high titers of neutralizing antibodies, which limits its clinical use. Here, we aim to establish a SAK mutant with equivalent activity to the wild type but reduced antigenicity, which may allow for multiple injections. Biosun software was used to predict SAK antigenic epitopes, and several main epitopes were modified by gene deletion and mutation. Ten SAK mutants were constructed, and their thrombolytic activity and immunogenicity were analyzed in vitro. SAK6, with a high expression level (45%), similar thrombolytic activity, and lower antibody reaction, was chosen for in vivo analysis in rhesus monkey. In the nearly 8-month experimental period, the antibody level of the SAK6 group was significantly lower than that of the SAK group. Moreover, only 5% of SAK activity was retained, whereas 75.6% of SAK6 activity was retained after incubating with respective antiserum. Overall, these results demonstrated that SAK6, established through comprehensive site-directed mutagenesis program, had identical thrombolytic activity to SAK, low immunogenicity, and less side effects, demonstrating its efficient clinical potential for thrombus disease.
Sigma-Aldrich
N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt, plasmin substrate