MilliporeSigma
  • Association between serum zinc and copper concentrations and copper/zinc ratio with the prevalence of knee chondrocalcinosis: a cross-sectional study.
Association between serum zinc and copper concentrations and copper/zinc ratio with the prevalence of knee chondrocalcinosis: a cross-sectional study.
BMC musculoskeletal disorders (2020-02-14)
Hongyi He, Yilun Wang, Zidan Yang, Xiang Ding, Tuo Yang, Guanghua Lei, Hui Li, Dongxing Xie
ABSTRACT
Patients with chondrocalcinosis may suffer from a series of symptoms resembling acute gouty arthritis or septic arthritis, but the aetiology and pathogenesis of chondrocalcinosis have not been fully elucidated yet. This study was aimed to assess serum zinc and copper concentrations, as well as the ratio of serum copper to zinc concentrations (Cu/Zn ratio), in relation to the prevalence of knee chondrocalcinosis. Data included in this analysis were retrieved from a large population-based cross-sectional study. A bilateral knee anteroposterior radiograph was obtained from each subject. Radiographic knee chondrocalcinosis was diagnosed if definite linear cartilage calcification was detected. Serum zinc and copper concentrations were measured using the spectrophotometric flow injection methods by Roche modular P800. The relations of serum zinc and copper concentrations and Cu/Zn ratio to the prevalence of knee chondrocalcinosis were examined using generalized estimating equations, respectively. The prevalence of knee chondrocalcinosis was 1.2% in the sample of this study (n = 12,362). In comparison with the lowest tertile, the odds ratios (ORs) of knee chondrocalcinosis adjusted by age, sex and body mass index were 0.74 (95% CI 0.50-1.09) in the second and 0.56 (95% CI 0.36-0.86) in the third tertiles of serum zinc concentrations (P for trend = 0.009), were 1.26 (95% CI 0.77-2.05) in the second and 2.01 (95% CI 1.25-3.24) in the third tertile of serum copper concentrations (P for trend = 0.003), and were 1.02 (95% CI 0.61-1.69) in the second and 2.23 (95% CI 1.38-3.59) in the third tertile of Cu/Zn ratio (P for trend < 0.001) respectively. These findings were not materially altered by adjustment for potential confounders. The present study observed that higher serum zinc concentrations, lower serum copper concentrations or lower Cu/Zn ratio are associated with a lower prevalence of knee chondrocalcinosis in a dose-response relationship manner.