MilliporeSigma
  • Overexpression of Sucrose Phosphate Synthase Enhanced Sucrose Content and Biomass Production in Transgenic Sugarcane.
Overexpression of Sucrose Phosphate Synthase Enhanced Sucrose Content and Biomass Production in Transgenic Sugarcane.
Plants (Basel, Switzerland) (2020-02-12)
Risky Mulana Anur, Nurul Mufithah, Widhi Dyah Sawitri, Hitoshi Sakakibara, Bambang Sugiharto
ABSTRACT
Sucrose phosphate synthase (SPS) is a key enzyme in sucrose synthesis, which controls sucrose content in plants. This study was designed to examine the efficacy of the overexpression of SoSPS1 gene on sucrose accumulation and carbon partitioning in transgenic sugarcane. The overexpression of SoSPS1 gene increased SPS activity and sucrose content in transgenic sugarcane leaves. More importantly, the overexpression enhanced soluble acid invertase (SAI) activity concomitant with the increase of glucose and fructose levels in the leaves, whereas sucrose synthase activity exhibited almost no change. In the stalk, a similar correlation was observed, but a higher correlation was noted between SPS activity and sugar content. These results suggest that SPS overexpression has both direct and indirect effects on sugar concentration and SAI activity in sugarcane. In addition, SPS overexpression resulted in a significant increase in plant height and stalk number in some transgenic lines compared to those in non-transgenic control. Taken together, these results strongly suggest that enhancing SPS activity is a useful strategy for improving sugarcane yield.
Product Description
GVS Transfer Membrane, Nitrocellulose, 0.45 um, W × L 30 cm × 3 m, roll of 1 roll