MilliporeSigma
  • Efficacy of human-simulated bronchopulmonary exposures of cefepime, zidebactam and the combination (WCK 5222) against MDR Pseudomonas aeruginosa in a neutropenic murine pneumonia model.
Efficacy of human-simulated bronchopulmonary exposures of cefepime, zidebactam and the combination (WCK 5222) against MDR Pseudomonas aeruginosa in a neutropenic murine pneumonia model.
The Journal of antimicrobial chemotherapy (2019-10-24)
James M Kidd, Kamilia Abdelraouf, David P Nicolau
ABSTRACT
WCK 5222 combines cefepime with zidebactam, a β-lactam enhancer that binds PBP2 and inhibits class A and C β-lactamases. The efficacy of human-simulated bronchopulmonary exposures of WCK 5222 against MDR Pseudomonas aeruginosa was investigated in a neutropenic murine pneumonia model. Nineteen MDR isolates of P. aeruginosa (cefepime MICs ≥64 mg/L) were studied. MICs of zidebactam and WCK 5222 ranged from 4 to 512 mg/L and from 4 to 32 mg/L, respectively. Dosing regimens of cefepime and zidebactam alone and in combination that achieved epithelial lining fluid (ELF) exposures in mice approximating human ELF exposures after doses of 2 g of cefepime/1 g of zidebactam every 8 h (1 h infusion) were utilized; controls were vehicle-dosed. Lungs were intranasally inoculated with 107-108 cfu/mL bacterial suspensions. Mice were dosed subcutaneously 2 h after inoculation for 24 h, then lungs were harvested. In vitro MIC was predictive of in vivo response to WCK 5222 treatment. Mean±SD changes in bacterial density at 24 h compared with 0 h controls (6.72±0.50 log10 cfu/lungs) for 13 isolates with WCK 5222 MICs ≤16 mg/L were 1.17±1.00, -0.99±1.45 and -2.21±0.79 log10 cfu/lungs for cefepime, zidebactam and WCK 5222, respectively. Against these isolates, zidebactam yielded >1 log10 cfu/lungs reductions in 8/13, while activity was enhanced with WCK 5222, producing >2 log10 cfu/lungs reductions in 10/13 and >1 log10 cfu/lungs reductions in 12/13. Among isolates with WCK 5222 MICs of 32 mg/L, five out of six showed a bacteriostatic response. Human-simulated bronchopulmonary exposure of WCK 5222 is effective against MDR P. aeruginosa at MIC ≤16 mg/L in a murine pneumonia model. These data support the clinical development of WCK 5222 for pseudomonal lung infections.
Sigma-Aldrich
5-Norbornene-2-endo,3-endo-dimethanol, 98%
Sigma-Aldrich
Mucin from porcine stomach, Type II