MilliporeSigma
  • Discovery of PROTAC BCL-XL degraders as potent anticancer agents with low on-target platelet toxicity.
Discovery of PROTAC BCL-XL degraders as potent anticancer agents with low on-target platelet toxicity.
European journal of medicinal chemistry (2020-03-08)
Xuan Zhang, Dinesh Thummuri, Xingui Liu, Wanyi Hu, Peiyi Zhang, Sajid Khan, Yaxia Yuan, Daohong Zhou, Guangrong Zheng
ABSTRACT
Anti-apoptotic protein BCL-XL plays a key role in tumorigenesis and cancer chemotherapy resistance, rendering it an attractive target for cancer treatment. However, BCL-XL inhibitors such as ABT-263 cannot be safely used in the clinic because platelets solely depend on BCL-XL to maintain their viability. To reduce the on-target platelet toxicity associated with the inhibition of BCL-XL, we designed and synthesized PROTAC BCL-XL degraders that recruit CRBN or VHL E3 ligase because both of these enzymes are poorly expressed in human platelets compared to various cancer cell lines. We confirmed that platelet-toxic BCL-XL/2 dual inhibitor ABT-263 can be converted into platelet-sparing CRBN/VHL-based BCL-XL specific degraders. A number of BCL-XL degraders are more potent in killing cancer cells than their parent compound ABT-263. Specifically, XZ739, a CRBN-dependent BCL-XL degrader, is 20-fold more potent than ABT-263 against MOLT-4 T-ALL cells and has >100-fold selectivity for MOLT-4 cells over human platelets. Our findings further demonstrated the utility of PROTAC technology to achieve tissue selectivity through recruiting differentially expressed E3 ligases.
Sigma-Aldrich
(S,R,S)-AHPC-Me, 95%
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder