MilliporeSigma
  • Improved biosynthesis of 5-hydroxymethyl-2-furancarboxylic acid and furoic acid from biomass-derived furans with high substrate tolerance of recombinant Escherichia coli HMFOMUT whole-cells.
Improved biosynthesis of 5-hydroxymethyl-2-furancarboxylic acid and furoic acid from biomass-derived furans with high substrate tolerance of recombinant Escherichia coli HMFOMUT whole-cells.
Bioresource technology (2020-02-11)
Zi-Wei Wang, Chun-Jie Gong, Yu-Cai He
ABSTRACT
The main aim of this work was to firstly develop a selective oxidation approach for biologically converting 5-hydroxymethylfurfural and furfural into the corresponding furan-based carboxylic acids with recombinant Escherichia coli HMFOMUT. Whole-cells of this recombinant strain harbored good biocatalytic activity in a narrow pH range (pH 6.5-7.0), which had high tolerance toward furfural (up to 50 mM) and 5-hydroxymethylfurfural (up to 150 mM), well-known potential inhibitors against microorganisms. 5-Hydroxymethyl-2-furancarboxylic acid and furoic acid could be obtained at 96.9% and 100% yield from 5-hydroxymethylfurfural (150 mM) and furfural (50 mM) at 30 °C and pH 7.0. The improved substrate tolerance of Escherichia coli HMFOMUT is gaining a great interest to synthesize value-added furan-based carboxylic acids, which has potential industrial applications.
Sigma-Aldrich
2-Furoic acid, 98%
Sigma-Aldrich
2,5-Furandicarboxaldehyde, 97%
Sigma-Aldrich
2,5-Furandicarboxylic acid, 97%