MilliporeSigma
  • Uranium(VI) Complexes of Glutathione Disulfide Forming in Aqueous Solution.
Uranium(VI) Complexes of Glutathione Disulfide Forming in Aqueous Solution.
Inorganic chemistry (2020-03-10)
Jerome Kretzschmar, Alexander Strobel, Toni Haubitz, Björn Drobot, Robin Steudtner, Astrid Barkleit, Vinzenz Brendler, Thorsten Stumpf
ABSTRACT
The interactions between glutathione disulfide, GSSG, the redox partner and dimer of the intracellular detoxification agent glutathione, GSH, and hexavalent uranium, U(VI), were extensively studied by solution NMR (in D2O), complemented by time-resolved laser-induced fluorescence and IR spectroscopies. As expected for the hard Lewis acid U(VI), coordination facilitates by the ligands' O-donor carboxyl groups. However, owing to the adjacent cationic α-amino group, the glutamyl-COO reveal monodentate binding, while the COO of the glycyl residues show bidentate coordination. The log K value for the reaction UO22+ + H3GSSG- → UO2(H3GSSG)+ (pH 3, 0.1 M NaClO4) was determined for the first time, being 4.81 ± 0.08; extrapolation to infinite dilution gave log K⊖ = 5.24 ± 0.08. U(VI) and GSSG form precipitates in the whole pD range studied (2-8), showing least solubility for 4 < pD < 6.5. Thus, particularly GSSG, hereby representing also other peptides and small proteins, affects the mobility of U(VI), strongly depending on the speciation of either component.
Sigma-Aldrich
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt, 97%