MilliporeSigma
  • Evaluation of phototoxicity of tattoo pigments using the 3 T3 neutral red uptake phototoxicity test and a 3D human reconstructed skin model.
Evaluation of phototoxicity of tattoo pigments using the 3 T3 neutral red uptake phototoxicity test and a 3D human reconstructed skin model.
Toxicology in vitro : an international journal published in association with BIBRA (2020-03-03)
So Young Kim, Sowoon Seo, Ki Hwan Choi, Jaesuk Yun
ABSTRACT
Phototoxicity due to dermally impregnated compounds exposed to ultraviolet radiation is associated with skin inflammation. The phototoxicity potential of active substances applied to the skin should be evaluated. Pigments are widely used for tattoos, and hypersensitivity reactions, such as photoallergic dermatitis, are possible tattoo-related complications. However, the phototoxicity of these chemicals is not well known. In this study, we evaluated the phototoxicity potential of six tattoo pigments, cadmium sulfide, carbazole, cadmium selenide, mercury (II) sulfide, chromium oxide, and cobalt aluminate, using in vitro methods-3 T3 neutral red uptake (NRU) phototoxicity test (PT) and a 3D human reconstructed skin model (EpiDerm). The validated 3 T3 NRU PT indicated the phototoxicity potential of carbazole and cadmium sulfide. The 3D human skin model confirmed that only carbazole was phototoxic. The 3 T3 NRU PT data corresponded well with those from the 3D skin model and suggested the need to employ several test systems for final phototoxicity assessment. In addition to the results obtained using 3 T3 NRU PT, further testing on 3D skin models may better reflect the bioavailability of a given chemical in the skin.
Sigma-Aldrich
Cobalt aluminum oxide, nanopowder, <50 nm particle size (BET), ≥99% trace metals basis (BET)