MilliporeSigma
Highlights of glucosamine-6P synthase catalysis.
Archives of biochemistry and biophysics (2008-02-19)
Philippe Durand, Béatrice Golinelli-Pimpaneau, Stéphane Mouilleron, Bernard Badet, Marie-Ange Badet-Denisot
ABSTRACT
L-Glutamine:d-fructose-6-phosphate amidotransferase, also known as glucosamine-6-phosphate synthase (GlcN6P synthase), which catalyzes the first step in a pathway leading to the formation of uridine 5'-diphospho-N-acetyl-d-glucosamine (UDP-GlcNAc), is a key point in the metabolic control of the biosynthesis of amino sugar-containing macromolecules. The molecular mechanism of the reaction catalyzed by GlcN6P synthase is complex and involves amide bond cleavage followed by ammonia channeling and sugar isomerization. This article provides a comprehensive overview of the present knowledge on this multi-faceted enzyme emphasizing the progress made during the last five years.
Sigma-Aldrich
D-Fructose 6-phosphate dipotassium salt, ≥97% (enzymatic), amorphous powder
Sigma-Aldrich
D-Fructose 6-phosphate disodium salt hydrate, ≥98%, amorphous powder