MilliporeSigma
  • PNU-282987 Attenuates Intestinal Epithelial Barrier Dysfunction in LPS-Induced Endotoxemia.
PNU-282987 Attenuates Intestinal Epithelial Barrier Dysfunction in LPS-Induced Endotoxemia.
Inflammation (2020-01-18)
Ying Zhang, Feng Zhou, Zhili Wang, Zhifeng Li, Jianguo Li
ABSTRACT
PNU-282987, the α7 acetylcholine receptor(α7nAchR) agonist, has been repeatedly reported to play a key role in anti-inflammatory action of multiple disease. However, little is known about its effect on LPS-induced intestinal epithelial barrier dysfunction. This study investigated the protective effects and mechanisms of PNU-282987 on intestinal epithelial barrier dysfunction in lipopolysaccharide(LPS)-induced endotoxemic rats. Endotoxemia models were induced by intraperitoneal injection of 10 mg/kg LPS. In the endotoxemic group, results showed increases in ileum mucosal permeability, ultrastructural damage of tight junction and redistribution of zonula occludens-1, apoptosis of intestinal epithelial cells and caspase-3 activation. These changes were significantly improved after PNU-282987 administration(P < 0.05). Pretreatment with α-bungarotoxin before PNU-282987 administration reversed the effects of PNU-282987(P < 0.05). These results indicate that PNU-282987 exerts protective effects on intestinal epithelial barrier dysfunction in LPS-induced endotoxemic rats, and its mechanism may involve the improvement of zonula occludens-1 and inhibition of enterocyte apoptosis in an α7nAchR-dependent manner.
Sigma-Aldrich
Sodium 4-hydroxybenzenesulfonate dihydrate, 98%