MilliporeSigma
  • 5-Hydroxymethylfurfural accumulation plays a critical role on acrylamide formation in coffee during roasting as confirmed by multiresponse kinetic modelling.
5-Hydroxymethylfurfural accumulation plays a critical role on acrylamide formation in coffee during roasting as confirmed by multiresponse kinetic modelling.
Food chemistry (2020-03-08)
Aytül Hamzalıoğlu, Vural Gökmen
ABSTRACT
This study aims to investigate in depth the mechanism of acrylamide formation in coffee during roasting. For this purpose, a comprehensive kinetic model including the elementary steps for acrylamide formation was proposed. The changes in sucrose, reducing sugars, free amino acids, asparagine, acrylamide, 3-deoxyglucosone, methylglyoxal, glyoxal, and 5-hydroxymethylfurfural were monitored in coffee during roasting at 200, 220 and 240 °C. Dominant pathways of complex reactions leading to acrylamide were unravelled by means of multiresponse kinetic modelling approach. The results of the model indicated that sucrose degrades into glucose and a reactive fructofuranosyl cation. Interestingly, glucose takes part mostly in the formation of intermediates, glyoxal and especially 3-deoxyglucosone rather than acrylamide formation. On the other hand, fructofuranosyl cation contributed mostly to the formation of 5-hydroxymethylfurfural which was found to be the most important intermediate precursor of acrylamide formed in coffee during roasting.
Sigma-Aldrich
3-Deoxyglucosone, ≥75% (TLC)
Sigma-Aldrich
Quinoxaline, ≥98%
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder