- Purification, crystallization and preliminary X-ray analysis of the glucosamine-6-phosphate N-acetyltransferase from human liver.
Purification, crystallization and preliminary X-ray analysis of the glucosamine-6-phosphate N-acetyltransferase from human liver.
Acta crystallographica. Section F, Structural biology and crystallization communications (2006-11-02)
Juan Wang, Yan-Feng Zhou, Lan-Fen Li, Yu-He Liang, Xiao-Dong Su
PMID17077487
ABSTRACT
Glucosamine-6-phosphate N-acetyltransferase from human liver, which catalyzes the transfer of an acetyl group from acetyl coenzyme A (AcCoA) to the primary amine of D-glucosamine 6-phosphate to form N-acetyl-D-glucosamine 6-phosphate, was expressed in a soluble form from Escherichia coli strain BL21 (DE3). The protein was purified to homogeneity using Ni(2+)-chelating chromatography followed by size-exclusion chromatography. Crystals of the protein were obtained by the hanging-drop vapour-diffusion method and diffracted to 2.6 A resolution. The crystals belonged to space group P4(1)2(1)2 or P4(3)2(1)2, with unit-cell parameters a = b = 50.08, c = 142.88 A.