MilliporeSigma
  • Effects of the CRF1 antagonist antalarmin on cocaine self-administration and discrimination in rhesus monkeys.
Effects of the CRF1 antagonist antalarmin on cocaine self-administration and discrimination in rhesus monkeys.
Pharmacology, biochemistry, and behavior (2006-12-22)
Nancy K Mello, S Stevens Negus, Kenner C Rice, Jack H Mendelson
ABSTRACT
Cocaine stimulates the rapid release of ACTH, and by inference, CRF in several species, suggesting that the HPA "stress" axis may contribute to the abuse-related effects of cocaine. The effects of a systemically-active CRF(1) receptor antagonist, antalarmin, on cocaine self-administration and cocaine discrimination were examined in rhesus monkeys. Antalarmin's acute (1-10 mg/kg, IV) and chronic (3.2 mg/kg IV) effects on IV cocaine self-administration were studied. The acute effects of 3.2 mg/kg IV antalarmin on the cocaine self-administration dose-effect curve (0.001-0.10 mg/kg/inj) were also examined. The acute effects of antalarmin (5 and 10 mg/kg, IM) on the cocaine discrimination dose-effect curve (0.013-1.3 mg/kg) were examined. Antalarmin did not significantly decrease the reinforcing or the discriminative stimulus effects of cocaine. Acute antalarmin administration produced a dose-dependent but non-significant decrease in self-administration of 0.01 mg/kg/inj cocaine but did not alter the cocaine dose-effect curve. Chronic daily antalarmin treatment did not significantly decrease cocaine-maintained responding. Antalarmin did not significantly alter either the cocaine discrimination dose-effect curve or the time course of the cocaine-training dose. Antalarmin (10 mg/kg) produced sedation, suggesting that it was centrally active, however, it did not attenuate cocaine's abuse-related effects in rhesus monkeys.
Sigma-Aldrich
Antalarmin hydrochloride, ≥98% (HPLC), solid