MilliporeSigma
  • Inference-Based Decisions in a Hidden State Foraging Task: Differential Contributions of Prefrontal Cortical Areas.
Inference-Based Decisions in a Hidden State Foraging Task: Differential Contributions of Prefrontal Cortical Areas.
Neuron (2020-02-13)
Pietro Vertechi, Eran Lottem, Dario Sarra, Beatriz Godinho, Isaac Treves, Tiago Quendera, Matthijs Nicolai Oude Lohuis, Zachary F Mainen
ABSTRACT
Essential features of the world are often hidden and must be inferred by constructing internal models based on indirect evidence. Here, to study the mechanisms of inference, we establish a foraging task that is naturalistic and easily learned yet can distinguish inference from simpler strategies such as the direct integration of sensory data. We show that both mice and humans learn a strategy consistent with optimal inference of a hidden state. However, humans acquire this strategy more than an order of magnitude faster than mice. Using optogenetics in mice, we show that orbitofrontal and anterior cingulate cortex inactivation impacts task performance, but only orbitofrontal inactivation reverts mice from an inference-based to a stimulus-bound decision strategy. These results establish a cross-species paradigm for studying the problem of inference-based decision making and begins to dissect the network of brain regions crucial for its performance.
Sigma-Aldrich
Gentamicin sulfate from Micromonospora purpurea, ≥590 μg/mg of gentamicin