MilliporeSigma
  • Effect of dapagliflozin, a sodium-glucose co-transporter-2 inhibitor, on gluconeogenesis in proximal renal tubules.
Effect of dapagliflozin, a sodium-glucose co-transporter-2 inhibitor, on gluconeogenesis in proximal renal tubules.
Diabetes, obesity & metabolism (2019-11-07)
Jin Hee Kim, Hae Young Ko, Hye Jin Wang, Hyangkyu Lee, MiJin Yun, Eun Seok Kang
ABSTRACT
To investigate the effect of dapagliflozin, a sodium-glucose co-transporter-2 (SGLT2) inhibitor, on renal gluconeogenesis in vitro, ex vivo and in vivo. We treated HK-2 cells (human renal proximal tubule cells) and mouse primary renal proximal tubule cells with dapagliflozin, and evaluated the process of renal gluconeogenesis. We also examined the effect of dapagliflozin on renal gluconeogenesis in normoglycaemic and hyperglycaemic mice. Dapagliflozin enhanced renal gluconeogenesis in vitro, ex vivo and in vivo. It increased phosphoenolpyruvate carboxykinase (PEPCK), glucose-6-phosphatase (G6Pase), peroxisome proliferative activated receptor-gamma co-activator 1α (PGC-1α) and phosphorylated cyclic-AMP response element binding protein (CREB) expression and decreased phosphorylated Forkhead Box O1 (FOXO1) expression in HK-2 cells, mouse primary renal proximal tubule cells, and the mouse renal cortex. Glutamine enhanced the gluconeogenic effect of dapagliflozin in HK-2 cells. Also, dapagliflozin increased 14 C-glutamine utilization in HK-2 cells. Glucagon did not affect dapagliflozin-induced enhancement in renal gluconeogenesis in HK-2 cells. SGLT2 gene knockdown with siRNA resulted in an increase of gluconeogenic gene expression and associated transcription factors in HK-2 cells. Dapagliflozin reduced fasting plasma glucose levels and improved oral glucose tolerance and insulin tolerance in high-fat diet-fed hyperglycaemic mice, although renal gluconeogenesis was enhanced. Dapagliflozin increased levels of gluconeogenic enzyme in the renal cortex and consequently increased renal gluconeogenesis, which is mediated by SGLT2 inhibition.
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC5A2
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)