MilliporeSigma
  • Inhibition of Proprotein Convertase Subtilisin/Kexin Type 9 Ameliorates Liver Fibrosis via Mitigation of Intestinal Endotoxemia.
Inhibition of Proprotein Convertase Subtilisin/Kexin Type 9 Ameliorates Liver Fibrosis via Mitigation of Intestinal Endotoxemia.
Inflammation (2019-11-30)
Yanting Zou, Shuyu Li, Beili Xu, Hongying Guo, Shuncai Zhang, Yu Cai
ABSTRACT
Lipopolysaccharide (LPS) is demonstrated to cause "two-hit" injury to liver. Proprotein convertase subtilisin/kexin type 9 (PCSK9) plays an important role in LPS clearance. Hepatocyte nuclear factor-1 alpha (HNF-1α) and sterol regulatory element-binding protein 2 (SREBP2) were reported to be responsible for PCSK9 gene transcription and regulation. We aim to clarify the expression status of PCSK9 during the process of liver fibrosis and to verify the effect on liver fibrosis via PCSK9 inhibition. In this study, we found that PCSK9 increased significantly in human and BDL mouse injured liver tissues, so did HNF-1α and SREBP2. No significant difference of plasma PCSK9 was observed. Inhibited PCSK9 using CRISPR-PCSK9 adeno-associated virus in BDL mice ameliorated liver inflammation and fibrosis, with LPS decrease in serum, without any change in intestinal wall integrity. PCSK9 expression of L02 hepatocytes can be induced by LPS; however, they lose the ability at high content of LPS. L02 cells increased LPS uptake after PCSK9 knockout. Taken together, these results suggest that, with PCSK9 increasing during liver fibrosis advancement, its inhibition can ameliorate liver injury by enhancing LPS uptake in hepatocytes; however, the enhancement is limited for destruction to hepatocytes by high LPS.
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
MISSION® esiRNA, targeting human PCSK9