MilliporeSigma
  • Characterization of the PEG layer of sterically stabilized liposomes: a SAXS study.
Characterization of the PEG layer of sterically stabilized liposomes: a SAXS study.
Chemistry and physics of lipids (2012-01-10)
Z Varga, A Wacha, U Vainio, J Gummel, A Bóta
ABSTRACT
Synchrotron small-angle X-ray scattering analysis of the bilayer structure of a pharmacologically relevant sterically stabilized liposome system is presented. Describing the electron density profile of the bilayer with the superposition of Gaussian functions, the contribution of the poly(ethylene glycol) (PEG) layers to the total electron density was identified. The changes in the thickness of the PEG layer as well as the distribution of the PEG chains among the outer and inner leaflets of the bilayers were followed by changing the molar ratio of the PEG-lipid and the molar weight of the PEG molecule.
Product Description
Avanti
18:0 PEG1000 PE, Avanti Research - A Croda Brand 880720C
Avanti
18:0 PEG1000 PE, Avanti Research - A Croda Brand 880720P, powder
Avanti
14:0 PEG1000 PE, Avanti Research - A Croda Brand 880710P, powder