MilliporeSigma
  • Selective knockdown of ceramide synthases reveals complex interregulation of sphingolipid metabolism.
Selective knockdown of ceramide synthases reveals complex interregulation of sphingolipid metabolism.
Journal of lipid research (2010-10-14)
Thomas D Mullen, Stefka Spassieva, Russell W Jenkins, Kazuyuki Kitatani, Jacek Bielawski, Yusuf A Hannun, Lina M Obeid
ABSTRACT
Mammalian ceramide synthases 1 to 6 (CerS1-6) generate Cer in an acyl-CoA-dependent manner, and expression of individual CerS has been shown to enhance the synthesis of ceramides with particular acyl chain lengths. However, the contribution of each CerS to steady-state levels of specific Cer species has not been evaluated. We investigated the knockdown of individual CerS in the MCF-7 human breast adenocarcinoma cell line by using small-interfering RNA (siRNA). We found that siRNA-induced downregulation of each CerS resulted in counter-regulation of nontargeted CerS. Additionally, each CerS knockdown produced unique effects on the levels of multiple sphingolipid species. For example, downregulation of CerS2 decreased very long-chain Cer but increased levels of CerS4, CerS5, and CerS6 expression and upregulated long-chain and medium-long-chain sphingolipids. Conversely, CerS6 knockdown decreased C16:0-Cer but increased CerS5 expression and caused non-C16:0 sphingolipids to be upregulated. Knockdown of individual CerS failed to decrease total sphingolipids or upregulate sphingoid bases. Treatment with siRNAs targeting combined CerS, CerS2, CerS5, and CerS6, did not change overall Cer or sphingomyelin mass but caused upregulation of dihydroceramide and hexosyl-ceramide and promoted endoplasmic reticulum stress. These data suggest that sphingolipid metabolism is robustly regulated by both redundancy in CerS-mediated Cer synthesis and counter-regulation of CerS expression.
Product Description
Avanti
C14 dihydroceramide (d18:0/14:0), Avanti Research - A Croda Brand 860632P, powder
Avanti
C18 Dihydroceramide (d18:0/18:0), Avanti Research - A Croda Brand 860627P, powder