MilliporeSigma
  • Sphingosine Kinase-2 Deficiency Ameliorates Kidney Fibrosis by Up-Regulating Smad7 in a Mouse Model of Unilateral Ureteral Obstruction.
Sphingosine Kinase-2 Deficiency Ameliorates Kidney Fibrosis by Up-Regulating Smad7 in a Mouse Model of Unilateral Ureteral Obstruction.
The American journal of pathology (2017-08-16)
Stephanie Schwalm, Sandra Beyer, Helena Frey, Riad Haceni, Georgios Grammatikos, Dominique Thomas, Gerd Geisslinger, Liliana Schaefer, Andrea Huwiler, Josef Pfeilschifter
ABSTRACT
Kidney fibrosis is a hallmark of chronic kidney disease and leads to extracellular matrix accumulation, organ scarring, and loss of kidney function. In this study, we investigated the role of sphingosine kinase-2 (SPHK2) on the progression of tubular fibrosis by using a mouse unilateral ureteral obstruction (UUO) model. We found that SPHK2 protein and activity are up-regulated in fibrotic renal tissue. Functionally, Sphk2-deficient (Sphk2-/-) mice showed an attenuated fibrotic response to UUO compared with wild-type mice, as demonstrated by reduced collagen abundance and decreased expression of fibronectin-1, collagen I, α-smooth muscle actin, connective tissue growth factor (CTGF), and plasminogen activator inhibitor (PAI-1). More important, these changes were associated with increased expression of the antifibrotic protein Smad7 and higher levels of sphingosine in Sphk2-/- UUO kidneys. Mechanistically, sphingosine ameliorates transforming growth factor-β-induced collagen accumulation, CTGF, and PAI-1 expression, but enhances Smad7 protein expression in primary kidney fibroblasts. In a complementary approach, in human Sphk2-overexpressing mice, UUO resulted in exacerbated signs of fibrosis with increased collagen accumulation, higher expression levels of fibronectin-1, collagen I, α-smooth muscle actin, CTGF, and PAI-1, but decreased Smad7 expression. SPHK2 plays an important role in kidney fibrogenesis by modulating transforming growth factor-β signaling. Thus, SPHK2 might be an attractive new target for the treatment of fibrosis in chronic kidney disease.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Avanti
sphingosine-d7, Avanti Research - A Croda Brand