MilliporeSigma
  • Lipid-PEG-folate encapsulated nanoparticles with aggregation induced emission characteristics: cellular uptake mechanism and two-photon fluorescence imaging.
Lipid-PEG-folate encapsulated nanoparticles with aggregation induced emission characteristics: cellular uptake mechanism and two-photon fluorescence imaging.
Small (Weinheim an der Bergstrasse, Germany) (2012-08-16)
Junlong Geng, Kai Li, Dan Ding, Xinhai Zhang, Wei Qin, Jianzhao Liu, Ben Zhong Tang, Bin Liu
ABSTRACT
Folate functionalized nanoparticles (NPs) that contain fluorogens with aggregation-induced emission (AIE) characteristics are fabricated to show bright far-red/near-infrared fluorescence, a large two-photon absorption cross section and low cytotoxicity, which are internalized into MCF-7 cancer cells mainly through caveolae-mediated endocytosis. One-photon excited in vivo fluorescence imaging illustrates that these AIE NPs can accumulate in a tumor and two-photon excited ex vivo tumor tissue imaging reveals that they can be easily detected in the tumor mass at a depth of 400 μm. These studies indicate that AIE NPs are promising alternatives to conventional TPA probes for biological imaging.
Product Description
Avanti
DSPE-PEG(5000) Folate, Avanti Research - A Croda Brand 880123X