MilliporeSigma
  • Plasma analysis of di- and trihydroxycholestanoic acid diastereoisomers in peroxisomal alpha-methylacyl-CoA racemase deficiency.
Plasma analysis of di- and trihydroxycholestanoic acid diastereoisomers in peroxisomal alpha-methylacyl-CoA racemase deficiency.
Journal of lipid research (2001-02-13)
S Ferdinandusse, H Overmars, S Denis, H R Waterham, R J Wanders, P Vreken
ABSTRACT
We identified a new peroxisomal disorder caused by a deficiency of the enzyme alpha-methylacyl-coenzyme A (CoA) racemase. Patients with this disorder show elevated plasma levels of pristanic acid and the bile acid intermediates di- and trihydroxycholestanoic acid (DHCA and THCA), which are all substrates for the peroxisomal beta-oxidation system. alpha-Methylacyl-CoA racemase plays an important role in the beta-oxidation of branched-chain fatty acids and fatty acid derivatives because it catalyzes the conversion of several (2R)-methyl-branched-chain fatty acyl-CoAs to their (2S)-isomers. Only stereoisomers with the 2-methyl group in the (S)-configuration can be degraded via beta-oxidation. In this study we used liquid chromatography/tandem mass spectrometry (LC-MS/MS) to analyze the bile acid intermediates that accumulate in plasma from patients with a deficiency of alpha-methylacyl-CoA racemase and, for comparison, in plasma from patients with Zellweger syndrome and patients with cholestatic liver disease.We found that racemase-deficient patients accumulate exclusively the (R)-isomer of free and taurine-conjugated DHCA and THCA, whereas in plasma of patients with Zellweger syndrome and patients with cholestatic liver disease both isomers were present. On the basis of these results we describe an easy and reliable method for the diagnosis of alpha-methylacyl-CoA racemase-deficient patients by plasma analysis. Our results also show that alpha-methylacyl-CoA racemase plays a unique role in bile acid formation. - Ferdinandusse, S., H. Overmars, S. Denis, H. R. Waterham, R. J. A. Wanders, and P. Vreken. Plasma analysis of di- and trihydroxycholestanoic acid diastereoisomers in peroxisomal alpha-methylacyl-CoA racemase deficiency. J. Lipid Res. 2001. 42: 137;-141.
Product Description
Avanti
trihydroxycholestanoic acid, Avanti Research - A Croda Brand