MilliporeSigma
  • 9-PAHSA promotes browning of white fat via activating G-protein-coupled receptor 120 and inhibiting lipopolysaccharide / NF-kappa B pathway.
9-PAHSA promotes browning of white fat via activating G-protein-coupled receptor 120 and inhibiting lipopolysaccharide / NF-kappa B pathway.
Biochemical and biophysical research communications (2018-10-21)
Yan-Mei Wang, Hong-Xia Liu, Ning-Yuan Fang
ABSTRACT
Browning of white adipose tissue is a novel mechanism to counteract obesity in view of its thermogenic activity. Activation of G-protein-coupled receptor 120 (GPR120) can promote the browning of white fat. 9-PAHSA, an endogenous mammalian lipid, which is acting as the ligand of GPR120 to enhance glucose uptake and exert anti-inflammatory effect. In the study, we would like to investigate the biological effects of 9-PAHSA on adipocyte browning. Here, we show that 9-PAHSA induces browning of 3T3-L1 adipocytes via enhanced expression of brown fat specific genes. 9-PAHSA-induced browning in white adipocytes of WT mice and ob/ob mice was investigated by determining expression levels of brown adipocyte-specific genes/proteins by quantitative real-time polymerase chain reaction analysis, immunoblot analysis and immunochemical staining. The effects of 9-PAHSA on brown fat markers in 3T3-L1 cells were decreased when GPR120 gene was silenced. To investigate the molecular mechanism of 9-PAHSA on adipocyte browning, lipopolysaccharide (LPS)-induced inflammatory model was conducted. 9-PAHSA treatment abolished LPS-induced NF-kappa B (NF-κB) activation and inflammatory cytokine secretion. But these anti-inflammatory effects of 9-PAHSA were attenuated by GPR120 knockdown. Our finding demonstrated that the browning of adipocyte was induced by 9-PAHSA through activating GPR120 and inhibiting the LPS/NF-κB pathway. This promising result will help to reveal the potential pathogenesis of obesity.
Product Description
Avanti
9-PAHSA, Avanti Research - A Croda Brand 900409C
Avanti
5-PAHSA, 5-(palmitoyloxy)octadecanoic acid, chloroform