MilliporeSigma
  • Metal cation induced cubic phase in poly(ethylene glycol)-functionalized dioleoylphosphatidylethanolamine aqueous dispersions.
Metal cation induced cubic phase in poly(ethylene glycol)-functionalized dioleoylphosphatidylethanolamine aqueous dispersions.
The journal of physical chemistry. B (2008-04-11)
Michela Pisani, Vincenzo Fino, Paolo Bruni, Emanuela Di Cola, Oriano Francescangeli
ABSTRACT
Metal cations (Mn(2+) or Ca(2+)) in aqueous dispersions of mixtures of dioleoylphosphatidylethanolamine (DOPE) and poly(ethylene glycol)-functionalized DOPE (DOPE-PEG(350)) induce, above a certain amount of the PEG lipid component, a phase transition from the inverted hexagonal phase H(II) to the bicontinuous inverted cubic phase Q(224) with space group Pn3m. The process is driven by the decrease of free elastic energy due to the Gaussian curvature of the cubic phase. The structural characterization of the phase behavior over the whole explored range of DOPE-PEG/DOPE weight ratio (3-25%) is reported, focusing on the role of the metal cation in the formation of the 3D cubic lattice. This result may represent a significant progress toward a design-based approach to drug delivery.
Product Description
Avanti
18:1 PEG350 PE, Avanti Research - A Croda Brand 880430O