MilliporeSigma
  • Generation and quality control of lipidomics data for the alzheimer's disease neuroimaging initiative cohort.
Generation and quality control of lipidomics data for the alzheimer's disease neuroimaging initiative cohort.
Scientific data (2018-11-21)
Dinesh Kumar Barupal, Sili Fan, Benjamin Wancewicz, Tomas Cajka, Michael Sa, Megan R Showalter, Rebecca Baillie, Jessica D Tenenbaum, Gregory Louie, Rima Kaddurah-Daouk, Oliver Fiehn
ABSTRACT
Alzheimer's disease (AD) is a major public health priority with a large socioeconomic burden and complex etiology. The Alzheimer Disease Metabolomics Consortium (ADMC) and the Alzheimer Disease Neuroimaging Initiative (ADNI) aim to gain new biological insights in the disease etiology. We report here an untargeted lipidomics of serum specimens of 806 subjects within the ADNI1 cohort (188 AD, 392 mild cognitive impairment and 226 cognitively normal subjects) along with 83 quality control samples. Lipids were detected and measured using an ultra-high-performance liquid chromatography quadruple/time-of-flight mass spectrometry (UHPLC-QTOF MS) instrument operated in both negative and positive electrospray ionization modes. The dataset includes a total 513 unique lipid species out of which 341 are known lipids. For over 95% of the detected lipids, a relative standard deviation of better than 20% was achieved in the quality control samples, indicating high technical reproducibility. Association modeling of this dataset and available clinical, metabolomics and drug-use data will provide novel insights into the AD etiology. These datasets are available at the ADNI repository at http://adni.loni.usc.edu/.
Product Description
Avanti
12:0-13:0 PC, Avanti Research - A Croda Brand LM1000, methanol solution
Avanti
17:0 SM (d18:1/17:0), Avanti Research - A Croda Brand 860585P, powder
Avanti
18:1-2:0 DG, 1-oleoyl-2-acetyl-sn-glycerol, chloroform