MilliporeSigma
  • Effects of oxiracetam on neurotransmitter release from rat hippocampus slices and synaptosomes.
Effects of oxiracetam on neurotransmitter release from rat hippocampus slices and synaptosomes.
Neuroscience letters (1992-09-28)
M Raiteri, R Costa, M Marchi
ABSTRACT
The effects of the nootropic drug oxiracetam on the K(+)-evoked overflow of [3H]D-aspartic acid ([3H]D-ASP), [3H]acetylcholine ([3H]ACh), [3H] gamma-aminobutyric acid ([3H]GABA), [3H]noradrenaline ([3H]NA) and [3H]5-hydroxytryptamine ([3H]5-HT) have been studied in superfused rat hippocampal slices. The overflow of [3H]D-ASP was enhanced by low concentrations of oxiracetam (0.01-1 microM) but not by high concentrations (10-100 microM) which showed some tendency to inhibit it. Similarly, low concentrations of oxiracetam increased, although less effectively, the depolarization-evoked overflow of [3H]ACh, whereas higher concentrations were without effect. At the concentrations active on [3H]D-ASP and [3H]ACh overflow oxiracetam did not affect that of [3H]GABA, [3H]NA or [3H]5-HT. The oxiracetam effects present in slices could not be observed in hippocampal synaptosomes. Thus oxiracetam may selectively increase the release of glutamate and acetylcholine in hippocampus by a mechanism which appears not to be sited in the releasing nerve terminals.