MilliporeSigma
  • Movement of zymosterol, a precursor of cholesterol, among three membranes in human fibroblasts.
Movement of zymosterol, a precursor of cholesterol, among three membranes in human fibroblasts.
The Journal of biological chemistry (1991-11-15)
Y Lange, F Echevarria, T L Steck
ABSTRACT
Where examined, cholesterol is synthesized in the endoplasmic reticulum; however, its precursor, zymosterol, is found mostly in the plasma membrane. The novel implication of these disparate findings is that zymosterol circulates within the cell. In tracing its movements, we have now established the following: (a) in human fibroblasts, zymosterol is converted to cholesterol solely in the rough ER. (b) Little or no zymosterol or cholesterol accumulates in the rough ER in vivo. (c) Newly synthesized zymosterol moves to the plasma membrane without a detectable lag and with a half-time of 9 min, about twice as fast as cholesterol. (d) The pool of radiolabeled zymosterol in the plasma membrane turns over rapidly, faster than does intracellular cholesterol. Thus, plasma membrane zymosterol is not stagnant. (e) [3H]Zymosterol pulsed into intact cells is initially found in the plasma membrane. It is rapidly internalized and is then converted to [3H] cholesterol. Half of the [3H]cholesterol produced returns to the plasma membrane within 30 min of the initial [3H]zymosterol pulse. (f) Nascent zymosterol accumulates in a buoyant sterol-rich intracellular membrane before it reaches the plasma membrane. This membrane also acquires nascent cholesterol, exogenous [3H]zymosterol pulsed into intact cells, and [3H]cholesterol synthesized from the exogenous [3H] zymosterol. These results suggest that at least one sterol moves rapidly and in both directions among the rough endoplasmic reticulum, a sterol-rich intracellular membrane bearing nascent cholesterol, and the plasma membrane.
Product Description
Avanti
zymosterol-d5, Avanti Research - A Croda Brand