MilliporeSigma
  • Synthesis and biological activities of 2'-deoxy-2'-fluoro-4'-thioarabinofuranosylpyrimidine and -purine nucleosides.
Synthesis and biological activities of 2'-deoxy-2'-fluoro-4'-thioarabinofuranosylpyrimidine and -purine nucleosides.
Bioorganic & medicinal chemistry (2000-09-08)
Y Yoshimura, K Kitano, K Yamada, S Sakata, S Miura, N Ashida, H Machida
ABSTRACT
As part of our ongoing investigation of the synthesis of biologically interesting 2'-modified-4'-thionucleosides, we synthesized 2'-deoxy-2'-fluoro-4'-thioarabinofuranosylpyrimidine and -purine nucleosides, and evaluated their antiviral and antitumor activities. In the pyrimidine series, beta-anomers of 5-ethyluracil, 5-iodouracil, 5-chloroethyluracil, and 5-iodocytosine derivatives showed potent and selective anti-HSV-1 and HSV-2 activities in vitro. In the purine series, guanine and 2,6-diaminopurine derivatives showed prominent antiviral activities with slight cytotoxicity. On the other hand, the 5-fluorocytosine derivative (5F-4'-thioFAC) showed potent antitumor activity against both leukemia and solid tumor. Its antitumor spectrum against 14 human solid tumor and one leukemic cell lines was compared with that of 4'-thioFAC. The results showed that 5F-4'-thioFAC had an antitumor spectrum similar to that of 4'-thioFAC. However, 5F-4'-thioFAC was about 10 times less active than 4'-thioFAC.
Sigma-Aldrich
5-Iodocytosine