MilliporeSigma
  • Investigation of stereoisomeric bisarylethenesulfonic acid esters for discovering potent and selective PTP1B inhibitors.
Investigation of stereoisomeric bisarylethenesulfonic acid esters for discovering potent and selective PTP1B inhibitors.
European journal of medicinal chemistry (2019-01-07)
Fangzhou Xie, Fengzhi Yang, Yaoyao Liang, Liang Li, Yu Xia, Faqin Jiang, Wenlu Liu, Yunyue Qi, Sharmin Reza Chowdhury, Dongsheng Xie, Lei Fu
ABSTRACT
Protein tyrosine phosphatase 1B (PTP1B) has been considered as a promising therapeutic target for type 2 diabetes mellitus (T2DM) and obesity due to its key regulating effects in insulin signaling and leptin receptor pathways. In this work, a series of cis- and trans-pyrrolidine bisarylethenesulfonic acid esters were prepared and their PTP1B inhibitory potency, selectivity and membrane permeability were evaluated. These novel stereoisomeric molecules especially trans-isomers exhibited remarkable inhibitory activity, significant selectivity as well as good membrane permeability (e.g. compound 28a, IC50 = 120, 1940 and 2670 nM against PTP1B, TCPTP and SHP2 respectively, and Papp = 1.74 × 10-6 cm/s). Molecular simulations indicated that trans-pyrrolidine bisarylethenesulfonic acid esters yielded the stronger binding affinity than their cis-isomers by constructing more interactions with non-catalytic sites of PTP1B. Further biological activity studies revealed that compound 28a could enhance insulin-stimulated glucose uptake and insulin-mediated insulin receptor β (IRβ) phosphorylation with no significant cytotoxicity.
Product Description
Avanti
PAMPA Lipid Blend I, Avanti Research - A Croda Brand