MilliporeSigma
Screening of Euphorbiaceae Plant Extracts for Anti-5α-reductase.
Biological & pharmaceutical bulletin (2018-08-03)
Hiroaki Kamei, Kazuma Noguchi, Hideaki Matsuda, Kazuya Murata
ABSTRACT
In our research program to find novel agents for alopecia from natural plant resources, we screened Euphorbiaceae plant extracts using an anti-5α-reductase assay. Among the samples tested, the extract of Phyllanthus urinaria showed the most potent activity with 24.3 and 64.6% inhibition at 50 and 200 µg/mL against the enzyme, respectively. The extract also suppressed the androgen activity of dihydrotestosterone in LNCaP cell line. These results show that the extract of P. urinaria may be a multi-potent agent for androgen-derived alopecia. We tested for activity on a hair regrowth model using mice. The extract of P. urinaria showed hair regrowth activity at 5 mg/mouse/d administration. Furthermore, the active principle for anti-5α-reductase activity was determined as stigmasterol glucoside from activity-guided fractionation and the IC50 was 27.2 µM. These results suggest that extract of P. urinaria may be a promising candidate anti-alopecia agent.
Product Description
Avanti
Glucosyl stigmasterol, Avanti Research - A Croda Brand 700160P, powder