MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Resolution, absolute stereochemistry, and enantiopharmacology of the GluR1-4 and GluR5 antagonist 2-amino-3-[5-tert-butyl-3-(phosphonomethoxy)-4-isoxazolyl]propionic acid.
Resolution, absolute stereochemistry, and enantiopharmacology of the GluR1-4 and GluR5 antagonist 2-amino-3-[5-tert-butyl-3-(phosphonomethoxy)-4-isoxazolyl]propionic acid.
Chirality (1999-11-24)
E H Møller, J Egebjerg, L Brehm, T B Stensbøl, T N Johansen, U Madsen, P Krogsgaard-Larsen
ABSTRACT
The phosphono amino acid, (RS)-2-amino-3-[5-tert-butyl-3-(phosphonomethoxy)-4-isoxazolyl+ ++]propio nic acid (ATPO), is a structural hybrid between the NMDA antagonist (RS)-2-amino-7-phosphonoheptanoic acid (AP7) and the AMPA and GluR5 agonist, (RS)-2-amino-3-(5-tert-butyl-3-hydroxy-4-isoxazolyl)propionic acid (ATPA). ATPO has been resolved into (S)-ATPO and (R)-ATPO using chiral HPLC, and the absolute stereochemistry of the two enantiomers was established by an X-ray crystallographic analysis of (R)-ATPO. (S)-ATPO and (R)-ATPO were characterized pharmacologically using rat brain membrane binding and electrophysiologically using the cortical wedge preparation as well as homo- or heteromeric GluR1-4, GluR5-6, and KA2 receptors expressed in Xenopus oocytes. (R)-ATPO was essentially inactive as an agonist or antagonist in all test systems. (S)-ATPO was an inhibitor of the binding of [(3)H]AMPA (IC(50) = 16 +/- 1 microM) and of [(3)H]-6-cyano-7-nitroquinoxaline-2,3-dione ([(3)H]CNQX) (IC(50) = 1.8 +/- 0.2 microM), but was inactive in the [(3)H]kainic acid and the [(3)H]-(RS)-3-(2-carboxypiperazin-4-yl)propyl-1-phosphonic acid ([(3)H]CPP) binding assays. (S)-ATPO did not show detectable agonist effects at any of the receptors under study, but antagonized AMPA-induced depolarization in the cortical wedge preparation (IC(50) = 15 +/- 1 microM). (S)-ATPO also blocked kainic acid agonist effects at GluR1 (K(i) = 2.0 microM), GluR1+2 (K(i) = 3.6 microM), GluR3 (K(i) = 3.6 microM), GluR4 (K(i) = 6.7 microM), and GluR5 (K(i) = 23 microM), but was inactive at GluR6 and GluR6+KA2. Thus, although ATPO is a structural analog of AP7 neither (S)-ATPO nor (R)-ATPO are recognized by NMDA receptor sites.