MilliporeSigma
  • Leveraging unique structural characteristics of WNK kinases to achieve therapeutic inhibition.
Leveraging unique structural characteristics of WNK kinases to achieve therapeutic inhibition.
Science signaling (2016-11-05)
Jinwei Zhang, Xianming Deng, Kristopher T Kahle
ABSTRACT
The with-no-lysine (K) WNK kinases are master regulators of the Na+-(K+)-Cl- cotransporters, including the renal-specific NCC and NKCC2 cotransporters. The discovery of WNK463, an orally bioavailable pan-WNK kinase inhibitor that exploits unique structural properties of the WNK catalytic domain to achieve high affinity and kinase selectivity, illustrates a strategy of leveraging distinct kinase features to develop specific inhibitors and validates the genetic predictions of the in vivo pharmacology of WNK inhibition.
Sigma-Aldrich
WNK463, ≥98% (HPLC)