MilliporeSigma
  • Toward the Rational Design of Universal Dual Polarity Matrix for MALDI Mass Spectrometry.
Toward the Rational Design of Universal Dual Polarity Matrix for MALDI Mass Spectrometry.
Analytical chemistry (2020-04-22)
Penghsuan Huang, Chun-Ying Huang, Ta-Chun Lin, Li-En Lin, Ethan Yang, Chuping Lee, Cheng-Chih Hsu, Pi-Tai Chou
ABSTRACT
A series of novel anthranilic acid derivatives I-IV, of which COOH-NH2 (I) and COOH-NHMe (IV) are endowed with acid and base bifunctionality, were designed and synthesized for matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry applications in dual polarity molecular imaging of biological samples, particularly for lipids. The heat of protonation, deprotonation, and proton transfer reaction as well as the capability of analyzing biomolecules in both positive and negative ion modes for I-IV were systematically investigated under standard 355 nm laser excitation. The results indicate correlation between dual polarity and acid-base property. Further, COOH-NHMe (IV) showed a unique performance and was successfully applied as the matrix for MALDI-TOF mass spectrometry imaging (MSI) for studying the mouse brain. Our results demonstrate the superiority of COOH-NHMe (IV) in detecting more lipid and protein species compared to commercially available matrices. Moreover, MALDI-TOF MSI results were obtained for lipid distributions, making COOH-NHMe (IV) a potential next generation universal matrix.
Product Description
Avanti
14:0 PS, Avanti Research - A Croda Brand
Avanti
14:0 PG, Avanti Research - A Croda Brand
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder
Avanti
14:0 PE, Avanti Research - A Croda Brand
Avanti
14:0 PA, 1,2-dimyristoyl-sn-glycero-3-phosphate (sodium salt), powder