MilliporeSigma
  • Differences in the effects of TRPV1 antagonists on energy metabolism in mice.
Differences in the effects of TRPV1 antagonists on energy metabolism in mice.
Biomedical research (Tokyo, Japan) (2018-12-12)
Jun Hai, Fuminori Kawabata, Yuko Kawabata, Ruojun Liang, Shotaro Nishimura, Shoji Tabata
ABSTRACT
Transient receptor potential vanilloid 1 (TRPV1) is a nociceptive cation channel that is activated by heat, protons and chemical ligands such as capsaicin. We investigated the roles of the capsaicin receptor, TRPV1, in controlling the energy metabolism of the whole body. It has been reported that the activation of TRPV1 by its agonists enhances energy metabolism. In this study, we used a respiratory gas analysis system to examine whether the inhibition of TRPV1 changes energy metabolism in mice. In addition, we examined the contributions of different modes of TRPV1 activation (heat, protons and capsaicin) to determine the influence of 3 different TRPV1 antagonists on energy metabolism. Here, we showed that intragastric administration of AMG517, a nonselective antagonist of TRPV1 (for heat, protons and capsaicin), enhanced energy metabolism as much as did intraperitoneal administration. On the other hand, intraperitoneal administration of AMG9810, a nonselective antagonist like AMG517, enhanced energy expenditure more than intragastric administration. However, the administration of JYL1421, a TRPV1 antagonist that very strongly inhibits TRPV1 activated by capsaicin, did not change energy metabolism. Taken together, these results suggest that the type of TRPV1 antagonists and the routes of its administration have different effects on energy metabolism in a normal body. Surprisingly, co-administration of JYL1421 and capsaicin significantly enhanced the energy metabolism more than administration of capsaicin alone. These results support the possibility that an unconventional mechanism is responsible for the increase in energy metabolism that occurs via TRPV1 inhibition.
Sigma-Aldrich
AMG517, ≥98% (HPLC)