MilliporeSigma
  • Discovery and characterization of natural products as novel indoleamine 2,3-dioxygenase 1 inhibitors through high-throughput screening.
Discovery and characterization of natural products as novel indoleamine 2,3-dioxygenase 1 inhibitors through high-throughput screening.
Acta pharmacologica Sinica (2019-06-15)
Wei Guo, Sheng Yao, Pu Sun, Tian-Biao Yang, Chun-Ping Tang, Ming-Yue Zheng, Yang Ye, Ling-Hua Meng
ABSTRACT
Indoleamine 2,3-dioxygenase 1 (IDO1) is emerging as a promising therapeutic target for the treatment of malignant tumors characterized by dysregulated tryptophan metabolism. However, the antitumor efficacy of existing small-molecule IDO1 inhibitors is still unsatisfactory, and the underlying mechanism remains largely undefined. To identify novel IDO1 inhibitors, an in-house natural product library of 2000 natural products was screened for inhibitory activity against recombinant human IDO1. High-throughput fluorescence-based screening identified 79 compounds with inhibitory activity > 30% at 20 μM. Nine natural products were further confirmed to inhibit IDO1 activity by > 30% using Ehrlich's reagent reaction. Compounds 2, 7, and 8 were demonstrated to inhibit IDO1 activity in a cellular context. Compounds 2 and 7 were more potent against IDO1 than TDO2 in the enzymatic assay. The kinetic studies showed that compound 2 exhibited noncompetitive inhibition, whereas compounds 7 and 8 were graphically well matched with uncompetitive inhibition. Compounds 7 and 8 were found to bind to the ferric-IDO1 enzyme. Docking stimulations showed that the naphthalene ring of compound 8 formed "T-shaped" π-π interactions with Phe-163 and that the 6-methyl-naphthalene group formed additional hydrophobic interactions with IDO1. Compound 8 was identified as a derivative of tanshinone, and preliminary SAR analysis indicated that tanshinone derivatives may be promising hits for the development of IDO1 inhibitors. This study provides new clues for the discovery of IDO1/TDO2 inhibitors with novel scaffolds.
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Methylene Blue hydrate, ≥95% (calc. to the dried substance)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils