MilliporeSigma
  • Generation of a homozygous HDAC6 knockout human embryonic stem cell line by CRISPR/Cas9 editing.
Generation of a homozygous HDAC6 knockout human embryonic stem cell line by CRISPR/Cas9 editing.
Stem cell research (2019-11-28)
Liangqun Xie, Jingrui Huang, Xun Li, Lei Dai, Xinxiu Lin, Jiejie Zhang, Jiefeng Luo, Weishe Zhang
ABSTRACT
Histone deacetylase 6 (HDAC6) is a unique cytoplasmic enzyme in the HDAC family. The HDAC6 has been shown to play important roles in several biological processes. Meanwhile, it is also an attractive therapeutic target for a variety of diseases. However, the mechanism of HDAC6 function is not fully understood yet, and it is still lacking highly specific targeted drugs. Here, we generated a homozygous HDAC6 knockout human embryonic stem cell (hESC) line, WAe009-A-21 by the CRISPR/Cas9-based gene editing method. The WAe009-A-21 cell line does not express HDAC6 protein, while maintaining normal 46, XX karyotype, pluripotency, and trilineage differentiation potential.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Wright stain, suitable for blood stain