MilliporeSigma
  • Search for clinical predictors of good glycemic control in patients starting or intensifying oral hypoglycemic pharmacological therapy: A multicenter prospective cohort study.
Search for clinical predictors of good glycemic control in patients starting or intensifying oral hypoglycemic pharmacological therapy: A multicenter prospective cohort study.
Journal of diabetes and its complications (2019-11-28)
Qian Ren, Li-Nong Ji, Ju-Ming Lu, Yu-Feng Li, Quan-Min Li, Shan-Shan Lin, Xiao-Feng Lv, Li Wang, Yuan Xu, Xiao-Hui Guo, Qi-Yu Guo, Li Ma, Jin Du, Ying-Li Chen, Cui-Ling Zhao, Qiu-Lan Zhang, Qi-Mei She, Xiu-Min Jiao, Mei-Hua Lu, Xiao-Meng Sun, Ying Gao, Jie Zhang
ABSTRACT
Our aim was to search for clinical predictors of good glycemic control in patients starting or intensifying oral hypoglycemic pharmacological therapy. A multicenter, prospective cohort of 499 diabetic subjects was enrolled in this study: patients with newly diagnosed diabetes (NDM group) or poor glycemic control with oral antidiabetic drugs (OADs) (PDM group). All subjects then started or intensified OADs therapy and followed up for 91 days. Glycemic control was determined according to HbA1c at day 91 with HbA1c <7% considered good. The proportions of patients with good glycemic control after follow up for 91 days were 66.9% and 34.8% in NDM group and PDM group respectively. Logistic regression analysis showed that the change in GA at 28 days was the only predictor of good glycemic control in NDM patients (OR = 1.630, 95% CI 1.300-2.044, P < 0.001). In PDM patients, changes in GA at 28 days, CPI, baseline HbA1c, diabetic duration, and BMI were all independent predictors of good glycemic control (All P < 0.05). GA decline is a good predictor of future success in newly diagnosed patients. In patients intensifying therapy, beside GA decline, other individualized clinical characteristics should also be considered.
Sigma-Aldrich
5-Norbornene-2-endo,3-endo-dimethanol, 98%