MilliporeSigma
microRNA-184 Induces a Commitment Switch to Epidermal Differentiation.
Stem cell reports (2017-12-05)
Sara Nagosa, Friederike Leesch, Daria Putin, Swarnabh Bhattacharya, Anna Altshuler, Laura Serror, Aya Amitai-Lange, Waseem Nasser, Edith Aberdam, Matthieu Rouleau, Sudhir G Tattikota, Matthew N Poy, Daniel Aberdam, Ruby Shalom-Feuerstein
ABSTRACT
miR-184 is a highly evolutionary conserved microRNA (miRNA) from fly to human. The importance of miR-184 was underscored by the discovery that point mutations in miR-184 gene led to corneal/lens blinding disease. However, miR-184-related function in vivo remained unclear. Here, we report that the miR-184 knockout mouse model displayed increased p63 expression in line with epidermal hyperplasia, while forced expression of miR-184 by stem/progenitor cells enhanced the Notch pathway and induced epidermal hypoplasia. In line, miR-184 reduced clonogenicity and accelerated differentiation of human epidermal cells. We showed that by directly repressing cytokeratin 15 (K15) and FIH1, miR-184 induces Notch activation and epidermal differentiation. The disease-causing miR-184C57U mutant failed to repress K15 and FIH1 and to induce Notch activation, suggesting a loss-of-function mechanism. Altogether, we propose that, by targeting K15 and FIH1, miR-184 regulates the transition from proliferation to early differentiation, while mis-expression or mutation in miR-184 results in impaired homeostasis.
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone LL002, clone LL002, Chemicon®, from mouse