MilliporeSigma
  • D-methionine (D-met) significantly rescues noise-induced hearing loss: timing studies.
D-methionine (D-met) significantly rescues noise-induced hearing loss: timing studies.
Hearing research (2011-09-20)
Kathleen Campbell, Alex Claussen, Robert Meech, Steven Verhulst, Daniel Fox, Larry Hughes
ABSTRACT
We have previously reported rescue from noise-induced auditory brainstem response (ABR) threshold shifts with d-methionine (d-met) administration 1 h after noise exposure. The present study investigated further d-met rescue intervals at 3, 5 and 7 h post-noise exposure. Chinchillas laniger were exposed to a 6 h 105 dB sound pressure level (dB SPL) octave band noise (OBN) and then administered d-met i.p. starting 3, 5, or 7 h after noise exposure; controls received saline i.p. immediately after noise exposure. ABR assessments were performed at baseline and on post-exposure days 1 and 21. Outer hair cell (OHC) loss was measured in cochleae obtained at sacrifice 21 days post-exposure. Administration of d-met starting at any of the delay times of 3-7 h post-noise exposure significantly reduced day 21 ABR threshold shift at 2 and 4 kHz and OHC loss at all hair cell regions measured (2, 4, 6 and 8 kHz). ABR threshold shifts in the control group at 6 and 8 kHz were only 8 and 11 dB respectively allowing little opportunity to observe protection at those 2 frequencies.
Sigma-Aldrich
D-Methionine, ≥98% (HPLC)