MilliporeSigma
  • Assemblies of Perisomatic GABAergic Neurons in the Developing Barrel Cortex.
Assemblies of Perisomatic GABAergic Neurons in the Developing Barrel Cortex.
Neuron (2019-11-30)
Laura Modol, Yannick Bollmann, Thomas Tressard, Agnès Baude, Alicia Che, Zhe Ran S Duan, Rachel Babij, Natalia V De Marco García, Rosa Cossart
ABSTRACT
The developmental journey of cortical interneurons encounters several activity-dependent milestones. During the early postnatal period in developing mice, GABAergic neurons are transient preferential recipients of thalamic inputs and undergo activity-dependent migration arrest, wiring, and programmed cell-death. Despite their importance for the emergence of sensory experience and the role of activity in their integration into cortical networks, the collective dynamics of GABAergic neurons during that neonatal period remain unknown. Here, we study coordinated activity in GABAergic cells of the mouse barrel cortex using in vivo calcium imaging. We uncover a transient structure in GABAergic population dynamics that disappears in a sensory-dependent process. Its building blocks are anatomically clustered GABAergic assemblies mostly composed by prospective parvalbumin-expressing cells. These progressively widen their territories until forming a uniform perisomatic GABAergic network. Such transient patterning of GABAergic activity is a functional scaffold that links the cortex to the external world prior to active exploration. VIDEO ABSTRACT.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-VGluT2 Antibody, serum, from guinea pig