MilliporeSigma
  • Characterizing smoking-induced transcriptional heterogeneity in the human bronchial epithelium at single-cell resolution.
Characterizing smoking-induced transcriptional heterogeneity in the human bronchial epithelium at single-cell resolution.
Science advances (2019-12-18)
Grant E Duclos, Vitor H Teixeira, Patrick Autissier, Yaron B Gesthalter, Marjan A Reinders-Luinge, Robert Terrano, Yves M Dumas, Gang Liu, Sarah A Mazzilli, Corry-Anke Brandsma, Maarten van den Berge, Sam M Janes, Wim Timens, Marc E Lenburg, Avrum Spira, Joshua D Campbell, Jennifer Beane
ABSTRACT
The human bronchial epithelium is composed of multiple distinct cell types that cooperate to defend against environmental insults. While studies have shown that smoking alters bronchial epithelial function and morphology, its precise effects on specific cell types and overall tissue composition are unclear. We used single-cell RNA sequencing to profile bronchial epithelial cells from six never and six current smokers. Unsupervised analyses led to the characterization of a set of toxin metabolism genes that localized to smoker ciliated cells, tissue remodeling associated with a loss of club cells and extensive goblet cell hyperplasia, and a previously unidentified peri-goblet epithelial subpopulation in smokers who expressed a marker of bronchial premalignant lesions. Our data demonstrate that smoke exposure drives a complex landscape of cellular alterations that may prime the human bronchial epithelium for disease.
Sigma-Aldrich
Anti-AKR1B10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MUC5B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid