MilliporeSigma
  • Novel, rapid, low-cost screen-printed (bio)sensors for the direct analysis of boar taint compounds androstenone and skatole in porcine adipose tissue: Comparison with a high-resolution gas chromatographic method.
Novel, rapid, low-cost screen-printed (bio)sensors for the direct analysis of boar taint compounds androstenone and skatole in porcine adipose tissue: Comparison with a high-resolution gas chromatographic method.
Biosensors & bioelectronics (2019-11-19)
K L Westmacott, A P Crew, O Doran, J P Hart
ABSTRACT
This is the first report on the fabrication, characterisation and application of an electrochemical (bio)sensor system for the simultaneous measurement of skatole and androstenone. A biosensor for androstenone was fabricated using a Meldola's Blue modified SPCE (MB-SPCE) by depositing NADH and the enzyme 3α-hydroxysteroid dehydrogenase onto the MB-SPCE surface; samples of adipose tissue were analysed using the biosensors in conjunction with chronoamperometry. Cyclic voltammetry was used to investigate the electrochemical behaviour of skatole at a screen-printed carbon electrode (SPCE vs. Ag/AgCl). An oxidation peak was observed around +0.55 V (vs. Ag/AgCl) and differential pulse voltammetry was applied for quantification of skatole in adipose tissue (in-situ). Quantitative analysis was achieved using calibration plots obtained from fortified meat samples. The concentrations obtained by the electrochemical and gas chromatographic (GC) methods demonstrated a good positive correlation. The (bio)sensor system completed both measurements within 60 s, as compared to several hours for GC, and at a considerably reduced cost and complexity. Consequently, the novel (bio)sensor system should have applications for analysis of carcasses on the abattoir processing line.
Sigma-Aldrich
5α-Androst-16-en-3-one
Sigma-Aldrich
5-Methylindole, 99%